1-(Benzyloxy)-4-fluoro-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

36750-25G 36750-5G
76888-610EA 364.95 USD
76888-610 76888-612
1-(Benzyloxy)-4-fluoro-2-methylbenzene ≥97%
1-(Benzyloxy)-4-fluoro-2-methylbenzene
Formula: C₁₄H₁₃FO
MW: 216.251 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24237546
CAS Number: 885267-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR