3,4-Dibromoiodobenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1,2-Dibromo-4-iodobenzene

36745-25G 36745-5G
76888-600EA 445.58 USD
76888-600 76888-602
3,4-Dibromoiodobenzene ≥97%
3,4-Dibromoiodobenzene

Precaución: for Research Use Only

Formula: C₆H₃Br₂I
Storage Temperature: Ambient
MDL Number: MFCD09753707
CAS Number: 909072-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR