1-(3-(Trifluoromethyl)phenyl)-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

36629-1G 36629-25G 36629-500MG 36629-5G
76888-544EA 142.17 USD
76888-544 76888-546 76888-548 76888-550
1-(3-(Trifluoromethyl)phenyl)-1H-pyrazole ≥97%
1-(3-(Trifluoromethyl)phenyl)-1H-pyrazole
Formula: C₁₀H₇F₃N₂
MW: 212.171 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12192635
CAS Number: 35715-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR