1-Bromo-2-(difluoromethyl)-4-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

36624-1G 36624-25G 36624-500MG 36624-5G
76888-534EA 126.25 USD
76888-534 76888-536 76888-538 76888-540
1-Bromo-2-(difluoromethyl)-4-methoxybenzene ≥95%
1-Bromo-2-(difluoromethyl)-4-methoxybenzene
Formula: C₈H₇BrF₂O
Storage Temperature: Refrigerator
MDL Number: MFCD14698634
CAS Number: 1214329-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR