1-Bromo-2,4-dichloro-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

36584-25G 36584-5G
76888-524EA 595.17 USD
76888-524 76888-526
1-Bromo-2,4-dichloro-3-methylbenzene ≥95%
1-Bromo-2,4-dichloro-3-methylbenzene
Formula: C₇H₅BrCl₂
MW: 239.93 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09878179
CAS Number: 127049-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR