1-Bromo-2-chloro-3,5-dimethoxybenzene ≥95%

Supplier: AOBChem USA

Warning

36542-1G 36542-25G 36542-500MG 36542-5G
76888-494EA 274.78 USD
76888-494 76888-496 76888-498 76888-500
1-Bromo-2-chloro-3,5-dimethoxybenzene ≥95%
1-Bromo-2-chloro-3,5-dimethoxybenzene
Formula: C₈H₈BrClO₂
Storage Temperature: Ambient
CAS Number: 81574-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR