2-(Difluoromethoxy)-3-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

36513-1G 36513-25G 36513-500MG 36513-5G
76888-482EA 173.99 USD
76888-482 76888-484 76888-486 76888-488
2-(Difluoromethoxy)-3-fluorobenzaldehyde ≥97%
2-(Difluoromethoxy)-3-fluorobenzaldehyde
Formula: C₈H₅F₃O₂
MW: 190.119 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14698498
CAS Number: 1214337-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR