2,4-Dibromo-5-methoxyaniline ≥97%

Supplier: AOBChem USA

Warning

36505-25G 36505-5G
76888-474EA 346.92 USD
76888-474 76888-476
2,4-Dibromo-5-methoxyaniline ≥97%
2,4-Dibromo-5-methoxyaniline
Formula: C₇H₇Br₂NO
Storage Temperature: Ambient
MDL Number: MFCD08282833
CAS Number: 35736-52-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR