2-(2-Ethoxynaphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

36446-25G 36446-5G
76888-416EA 471.04 USD
76888-416 76888-418
2-(2-Ethoxynaphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(2-Ethoxynaphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₈H₂₃BO₃
MW: 298.184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632015
CAS Number: 2246703-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR