1-Bromo-2-(difluoromethoxy)-3-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

36389-1G 36389-25G 36389-500MG 36389-5G
76888-400EA 123.07 USD
76888-400 76888-402 76888-404 76888-406
1-Bromo-2-(difluoromethoxy)-3-fluorobenzene ≥97%
1-Bromo-2-(difluoromethoxy)-3-fluorobenzene
Formula: C₇H₄BrF₃O
Storage Temperature: Refrigerator
MDL Number: MFCD11036073
CAS Number: 954235-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR