1-Bromo-3-(methoxymethyl)-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

36336-1G 36336-25G 36336-500MG 36336-5G
76888-362EA 155.96 USD
76888-362 76888-364 76888-366 76888-368
1-Bromo-3-(methoxymethyl)-2-methylbenzene ≥97%
1-Bromo-3-(methoxymethyl)-2-methylbenzene
Formula: C₉H₁₁BrO
MW: 215.087 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32631998
CAS Number: 2169386-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR