2,2,2-Trifluoroacetophenone-4-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,2,2-Trifluoro-1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethanone

36311-1G 36311-25G 36311-500MG 36311-5G
76888-320EA 159.14 USD
76888-320 76888-322 76888-324 76888-326
2,2,2-Trifluoroacetophenone-4-boronic acid pinacol ester ≥97%
2,2,2-Trifluoroacetophenone-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₆BF₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD16628195
CAS Number: 1004294-77-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR