2-(3-Bromophenyl)-1H-benzo[d]imidazole ≥97%

Supplier: AOBChem USA

Warning

36298-1G 36298-25G 36298-500MG 36298-5G
76888-308EA 203.7 USD
76888-308 76888-310 76888-312 76888-314
2-(3-Bromophenyl)-1H-benzo[d]imidazole ≥97%
2-(3-Bromophenyl)-1H-benzo[d]imidazole
Formula: C₁₃H₉BrN₂
Storage Temperature: Refrigerator
MDL Number: MFCD00602186
CAS Number: 77738-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR