2-(4-Bromophenyl)benzimidazole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(4-Bromophenyl)-1H-benzo[d]imidazole

36284-1G 36284-25G 36284-5G
76888-290EA 76.39 USD
76888-290 76888-292 76888-294
2-(4-Bromophenyl)benzimidazole ≥97%
2-(4-Bromophenyl)benzimidazole
Formula: C₁₃H₉BrN₂
MW: 273.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00224360
CAS Number: 2622-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR