1,2-Difluoro-3-(methoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

36238-1G 36238-25G 36238-5G
76888-270EA 62.6 USD
76888-270 76888-272 76888-274
1,2-Difluoro-3-(methoxymethyl)benzene ≥97%
1,2-Difluoro-3-(methoxymethyl)benzene
Formula: C₈H₈F₂O
MW: 158.145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631982
CAS Number: 865664-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR