1-(6-Bromopyridin-3-yl)-2,2,2-trifluoroethanol ≥97%

Supplier: AOBChem USA

Warning

36193-1G 36193-25G 36193-500MG 36193-5G
76888-226EA 379.81 USD
76888-226 76888-228 76888-230 76888-232
1-(6-Bromopyridin-3-yl)-2,2,2-trifluoroethanol ≥97%
1-(6-Bromopyridin-3-yl)-2,2,2-trifluoroethanol
Formula: C₇H₅BrF₃NO
Storage Temperature: Refrigerator
MDL Number: MFCD21604764
CAS Number: 1188477-81-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR