1-Bromo-2,5-dimethoxy-4-(methoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

36129-1G 36129-500MG 36129-5G
76888-214EA 689.59 USD
76888-214 76888-216 76888-218
1-Bromo-2,5-dimethoxy-4-(methoxymethyl)benzene ≥97%
1-Bromo-2,5-dimethoxy-4-(methoxymethyl)benzene
Formula: C₁₀H₁₃BrO₃
Storage Temperature: Refrigerator
MDL Number: MFCD32631956
CAS Number: 2379321-66-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR