(3-Chloro-4-(difluoromethoxy)phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

36025-1G 36025-500MG 36025-5G 36025-25G
76888-172EA 561.22 USD
76888-172 76888-174 76888-176 77655-396
(3-Chloro-4-(difluoromethoxy)phenyl)methanol ≥95%
(3-Chloro-4-(difluoromethoxy)phenyl)methanol
Formula: C₈H₇ClF₂O₂
MW: 208.59 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16075369
CAS Number: 1249809-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR