1-Bromo-4,5-dichloro-2-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

35905-25G 35905-5G
76888-162EA 194.15 USD
76888-162 76888-164
1-Bromo-4,5-dichloro-2-nitrobenzene ≥97%
1-Bromo-4,5-dichloro-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrCl₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD07780670
CAS Number: 93361-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR