2,2,2-Trifluoro-1-(p-tolyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

35870-1G 35870-25G 35870-5G
76888-148EA 66.84 USD
76888-148 76888-150 76888-152
2,2,2-Trifluoro-1-(p-tolyl)ethanol ≥97%
2,2,2-Trifluoro-1-(p-tolyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₉F₃O
Storage Temperature: Ambient
MDL Number: MFCD00233111
CAS Number: 446-65-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR