4'-Chloro-3'-methylacetophenone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-(4-Chloro-3-methylphenyl)ethanone

35841-25G 35841-5G
76888-130EA 183.54 USD
76888-130 76888-132
4'-Chloro-3'-methylacetophenone ≥97%
4'-Chloro-3'-methylacetophenone

Precaución: for Research Use Only

Formula: C₉H₉ClO
MW: 168.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00045172
CAS Number: 37074-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR