2-(Methoxymethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

35760-1G 35760-25G 35760-500MG 35760-5G
76888-084EA 361.77 USD
76888-084 76888-086 76888-088 76888-090
2-(Methoxymethyl)benzaldehyde ≥97%
2-(Methoxymethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
Storage Temperature: Refrigerator
MDL Number: MFCD24395542
CAS Number: 106020-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR