1-tert-Butyl-1H-pyrazole ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
1-(tert-Butyl)-1H-pyrazole
35701-25G
35701-5G
76888-072EA
219.61
USD
76888-072
76888-074
1-tert-Butyl-1H-pyrazole ≥97%
1-tert-Butyl-1H-pyrazole
Precaución: for Research Use Only
Formula:
C₇H₁₂N₂ Storage Temperature: Ambient |
MDL Number:
MFCD12192641 CAS Number: 15754-60-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...