1-Acetyl-4-fluoronaphthalene ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4'-Fluoro-1'-acetonaphthone, 1-(4-Fluoronaphthalen-1-yl)ethanone, 1-(4-Fluoro-1-naphthyl)ethanone

35569-25G 35569-5G
76888-062EA 233.4 USD
76888-062 76888-064
1-Acetyl-4-fluoronaphthalene ≥97% research grade
1-Acetyl-4-fluoronaphthalene
Formula: C₁₂H₉FO
MW: 188.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00134475
CAS Number: 316-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR