(6-Cyano-2-methylpyridin-3-yl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

35441-1G 35441-500MG 35441-5G
76888-046EA 968.61 USD
76888-046 76888-048 76888-050
(6-Cyano-2-methylpyridin-3-yl)boronic acid ≥95%
(6-Cyano-2-methylpyridin-3-yl)boronic acid
Formula: C₇H₇BN₂O₂
MW: 161.954 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22491542
CAS Number: 2225180-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR