1-(4-Chloro-3-ethoxyphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

35416-1G 35416-25G 35416-5G
76888-032EA 108.22 USD
76888-032 76888-034 76888-036
1-(4-Chloro-3-ethoxyphenyl)ethanone ≥97%
1-(4-Chloro-3-ethoxyphenyl)ethanone
Formula: C₉H₉ClO₃
Storage Temperature: Ambient
CAS Number: 97209-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR