2-(Benzyloxy)-4-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

35271-1G 35271-500MG 35271-5G
76888-008EA 601.54 USD
76888-008 76888-010 76888-012
2-(Benzyloxy)-4-methylphenylboronic acid ≥97%
2-(Benzyloxy)-4-methylphenylboronic acid
Formula: C₁₄H₁₅BO₃
MW: 242.078 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18383549
CAS Number: 2246603-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR