3-Bromo-2-fluoro-6-iodotoluene ≥97%

Supplier: AOBChem USA

Warning

35100-1G 35100-25G 35100-5G 35100-500MG
76887-990EA 96.55 USD
76887-990 76887-992 76887-994 77646-274
3-Bromo-2-fluoro-6-iodotoluene ≥97%
3-Bromo-2-fluoro-6-iodotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrFI
Storage Temperature: Refrigerator
MDL Number: MFCD09800764
CAS Number: 1000576-29-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR