(5-Bromo-2-Chlorophenyl)(Methyl)Sulfane ≥97%

Supplier: AOBChem USA

Warning

35058-1G 35058-25G 35058-5G 35058-500MG
76887-978EA 111.4 USD
76887-978 76887-980 76887-982 77646-272
(5-Bromo-2-Chlorophenyl)(Methyl)Sulfane ≥97%
(5-Bromo-2-Chlorophenyl)(Methyl)Sulfane
Formula: C₇H₆BrClS
MW: 237.545 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22488922
CAS Number: 740806-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR