(2-Fluoro-6-methoxy-4-methylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

34903-1G 34903-500MG 34903-5G
76887-966EA 683.22 USD
76887-966 76887-968 76887-970
(2-Fluoro-6-methoxy-4-methylphenyl)methanol ≥95%
(2-Fluoro-6-methoxy-4-methylphenyl)methanol
Formula: C₉H₁₁FO₂
MW: 170.181 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31805669
CAS Number: 2229187-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR