2,3-Difluoro-6-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3,4-Difluor-2-hydroxyanisole

34821-1G 34821-25G 34821-5G 34821-500MG
76887-936EA 79.57 USD
76887-936 76887-938 76887-940 77646-252
2,3-Difluoro-6-methoxyphenol ≥97%
2,3-Difluoro-6-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₆F₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD04116024
CAS Number: 186306-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR