1-(2-Bromo-6-fluorophenyl)-N,N-dimethylmethanamine ≥97%

Supplier: AOBChem USA

Warning

34804-1G 34804-25G 34804-500MG 34804-5G
76887-924EA 137.92 USD
76887-924 76887-926 76887-928 76887-930
1-(2-Bromo-6-fluorophenyl)-N,N-dimethylmethanamine ≥97%
1-(2-Bromo-6-fluorophenyl)-N,N-dimethylmethanamine
Formula: C₉H₁₁BrFN
MW: 232.093 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21333061
CAS Number: 1355247-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR