1,3-Dimethyl-5-fluoro-2-iodobenzene 95%
Supplier: AOBChem USA
|
Warning
|
34775-1G
34775-25G
34775-5G
34775-500MG
76887-910EA
112.46
USD
76887-910
76887-912
76887-914
77646-250
1,3-Dimethyl-5-fluoro-2-iodobenzene 95%
1,3-Dimethyl-5-fluoro-2-iodobenzene
Formula:
C₈H₈FI MW: 250.052 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD28016865 CAS Number: 14659-59-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...