2,3-Difluoro-1,4-benzenediol ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2,3-Difluorohydroquinone

34668-1G 34668-25G 34668-500MG 34668-5G
76887-898EA 163.38 USD
76887-898 76887-900 76887-902 76887-904
2,3-Difluoro-1,4-benzenediol ≥97% research grade
2,3-Difluoro-1,4-benzenediol
Formula: C₆H₄F₂O₂
MW: 146.092 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04038791
CAS Number: 124728-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR