3',5'-Dichloro-2,2,2-trifluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-(3,5-Dichlorophenyl)-2,2,2-trifluoroethanone

34410-25G 34410-5G
76887-860EA 418 USD
76887-860 76887-862
3',5'-Dichloro-2,2,2-trifluoroacetophenone ≥97%
3',5'-Dichloro-2,2,2-trifluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₃Cl₂F₃O
MW: 243.0100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01319996
CAS Number: 130336-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR