2,3-Dibromo-6-chloropyridine ≥97%

Supplier: AOBChem USA

Warning

34355-25G 34355-5G
76887-838EA 407.39 USD
76887-838 76887-840
2,3-Dibromo-6-chloropyridine ≥97%
2,3-Dibromo-6-chloropyridine

Precaución: for Research Use Only

Formula: C₅H₄ClN
MW: 113.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06798229
CAS Number: 885952-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR