4-Fluoro-3-iodobenzotrifluoride ≥97%
Supplier: AOBChem USA
|
Warning
|
34334-25G
34334-5G
76887-826EA
516.66
USD
76887-826
76887-828
4-Fluoro-3-iodobenzotrifluoride ≥97%
4-Fluoro-3-iodobenzotrifluoride
Precaución: for Research Use Only
Formula:
C₇H₃F₄I Storage Temperature: Ambient |
MDL Number:
MFCD09800715 CAS Number: 110192-48-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...