1-Chloro-3-ethoxy-2-fluorobenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
34106-1G
34106-25G
34106-5G
76887-768EA
111.4
USD
76887-768
76887-770
76887-772
1-Chloro-3-ethoxy-2-fluorobenzene ≥97%
1-Chloro-3-ethoxy-2-fluorobenzene
Formula:
C₈H₈ClFO Storage Temperature: Ambient |
MDL Number:
MFCD27932108 CAS Number: 909122-16-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...