1-Bromo-2-chloro-3-fluoro-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

34104-1G 34104-25G 34104-500MG 34104-5G
76887-758EA 272.66 USD
76887-758 76887-760 76887-762 76887-764
1-Bromo-2-chloro-3-fluoro-4-methoxybenzene ≥97%
1-Bromo-2-chloro-3-fluoro-4-methoxybenzene
Formula: C₇H₅BrClFO
Storage Temperature: Refrigerator
MDL Number: MFCD19440931
CAS Number: 909122-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR