2,3-Dichloro-4-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

34076-1G 34076-25G 34076-500MG 34076-5G
76887-742EA 296 USD
76887-742 76887-744 76887-746 76887-748
2,3-Dichloro-4-methoxyphenylboronic acid ≥97%
2,3-Dichloro-4-methoxyphenylboronic acid
Formula: C₇H₇BCl₂O₃
Storage Temperature: Ambient
CAS Number: 1190219-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR