1-Bromo-3-chloro-4-fluoro-2-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

34023-1G 34023-25G 34023-500MG 34023-5G
76887-726EA 264.17 USD
76887-726 76887-728 76887-730 76887-732
1-Bromo-3-chloro-4-fluoro-2-methoxybenzene ≥95%
1-Bromo-3-chloro-4-fluoro-2-methoxybenzene
Formula: C₇H₅BrClFO
MW: 239.469 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29044048
CAS Number: 1784628-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR