(6-Chloro-5-methylpyridin-3-yl)methylamine hydrochloride ≥97%

Supplier: AOBChem USA

Warning

33741-1G 33741-25G 33741-500MG 33741-5G
76887-682EA 260.99 USD
76887-682 76887-684 76887-686 76887-688
(6-Chloro-5-methylpyridin-3-yl)methylamine hydrochloride ≥97%
(6-Chloro-5-methylpyridin-3-yl)methylamine hydrochloride
Formula: C₇H₁₀Cl₂N₂
MW: 193.074 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17214193
CAS Number: 1257535-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR