(6-Bromo-2-fluoro-3-(trifluoromethoxy)phenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

33662-1G 33662-25G 33662-500MG 33662-5G
76887-656EA 337.37 USD
76887-656 76887-658 76887-660 76887-662
(6-Bromo-2-fluoro-3-(trifluoromethoxy)phenyl)boronic acid ≥97%
(6-Bromo-2-fluoro-3-(trifluoromethoxy)phenyl)boronic acid
Formula: C₇H₄BBrF₄O₃
MW: 302.813 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32196872
CAS Number: 2376634-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR