1-Bromo-3-Fluoro-4-Methyl-2-(Methylsulfanyl)Benzene ≥97%

Supplier: AOBChem USA

Warning

33639-25G 33639-5G
76887-644EA 310.85 USD
76887-644 76887-646
1-Bromo-3-Fluoro-4-Methyl-2-(Methylsulfanyl)Benzene ≥97%
1-Bromo-3-Fluoro-4-Methyl-2-(Methylsulfanyl)Benzene
Formula: C₈H₈BrFS
Storage Temperature: Ambient
MDL Number: MFCD31382562
CAS Number: 2121513-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR