1,3-Dibromo-5-pentoxybenzene ≥97%

Supplier: AOBChem USA

Warning

33579-25G 33579-5G
76887-620EA 519.85 USD
76887-620 76887-622
1,3-Dibromo-5-pentoxybenzene ≥97%
1,3-Dibromo-5-pentoxybenzene
Formula: C₁₁H₁₄Br₂O
MW: 322.036 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30834082
CAS Number: 2121515-32-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR