2,3-Dimethyl-4-(phenylmethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

33337-1G 33337-25G 33337-500MG 33337-5G
76887-588EA 171.87 USD
76887-588 76887-590 76887-592 76887-594
2,3-Dimethyl-4-(phenylmethoxy)benzaldehyde ≥97%
Formula: C₁₆H₁₆O₂
Storage Temperature: Refrigerator
MDL Number: MFCD30834045
CAS Number: 1609289-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR