[4-Fluoro-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanol ≥97%

Supplier: AOBChem USA

Warning

33130-1G 33130-25G 33130-500MG 33130-5G
76887-542EA 153.84 USD
76887-542 76887-544 76887-546 76887-548
[4-Fluoro-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanol ≥97%
[4-Fluoro-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanol
Formula: C₁₃H₁₈BFO₃
MW: 252.0900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18729956
CAS Number: 1333264-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR