[2-(Benzyloxy)-4-bromophenyl]methanol ≥97%

Supplier: AOBChem USA

Warning

32969-1G 32969-25G 32969-5G
76887-528EA 64.72 USD
76887-528 76887-530 76887-532
[2-(Benzyloxy)-4-bromophenyl]methanol ≥97%
[2-(Benzyloxy)-4-bromophenyl]methanol
Formula: C₁₄H₁₃BrO₂
MW: 293.156 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11651997
CAS Number: 1094474-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR