2-(Benzyloxy)-3-chlorobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

32956-1G 32956-25G 32956-500MG 32956-5G
76887-514EA 225.98 USD
76887-514 76887-516 76887-518 76887-520
2-(Benzyloxy)-3-chlorobenzaldehyde ≥95%
2-(Benzyloxy)-3-chlorobenzaldehyde
Formula: C₁₄H₁₁ClO₂
Storage Temperature: Ambient
MDL Number: MFCD26056350
CAS Number: 80352-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR