1-Bromo-4-[(Difluoromethyl)Sulfanyl]Benzene ≥97% research grade

Supplier: AOBChem USA

Warning

32752-1G 32752-25G 32752-500MG 32752-5G
76887-472EA 177.18 USD
76887-472 76887-474 76887-476 76887-478
1-Bromo-4-[(Difluoromethyl)Sulfanyl]Benzene ≥97% research grade
1-Bromo-4-[(Difluoromethyl)Sulfanyl]Benzene
Formula: C₇H₅BrF₂S
Storage Temperature: Refrigerator
MDL Number: MFCD00766771
CAS Number: 4837-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR